sec-Butyl chloroformate 95%

Supplier: Apollo Scientific

OR917261-1G OR917261-25G OR917261-5G
APOSOR917261-1GEA 145 EUR
APOSOR917261-1G APOSOR917261-25G APOSOR917261-5G
sec-Butyl chloroformate 95%
sec-Butyl chloroformate
Formula: C₅H₉ClO₂
MW: 136,577 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00043796
CAS Number: 17462-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR