5-Hydroxy-4-methyl-2(5H)furanone 95%
Supplier: Apollo Scientific
Synonyms:
5-Hydroxy-4-methylfuran-2(5H)-one
OR917392-1G
OR917392-5G
OR917392-25G
APOSOR917392-1GEA
58.2
EUR
APOSOR917392-1G
APOSOR917392-5G
APOSOR917392-25G
5-Hydroxy-4-methyl-2(5H)furanone 95%
5-Hydroxy-4-methyl-2(5H)furanone
Formula:
C₅H₆O₃ MW: 114,099 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07782086 CAS Number: 40834-42-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...