1-Allyl-1H-benzimidazole-2-carbaldehyde 95%
Supplier: Apollo Scientific
OR917409-1G
OR917409-250MG
APOSOR917409-1GEA
401
EUR
APOSOR917409-1G
APOSOR917409-250MG
1-Allyl-1H-benzimidazole-2-carbaldehyde 95%
1-Allyl-1H-benzimidazole-2-carbaldehyde
Formula:
C₁₁H₁₀N₂O MW: 186,21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04355428 CAS Number: 118482-14-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...