2-Methyl-5,6,7,8-tetrahydroquinazolin-4-ol 95%

Supplier: Apollo Scientific

OR917525-1G OR917525-5G
APOSOR917525-1GEA 104 EUR
APOSOR917525-1G APOSOR917525-5G
2-Methyl-5,6,7,8-tetrahydroquinazolin-4-ol 95%
2-Methyl-5,6,7,8-tetrahydroquinazolin-4-ol
Formula: C₉H₁₂N₂O
MW: 164,204 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16872923
CAS Number: 19178-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR