4-Methyl-2-oxo-2H-chromene-7-carbaldehyde 95%
Supplier: Apollo Scientific
OR917609-1G
OR917609-250MG
APOSOR917609-1GEA
713
EUR
APOSOR917609-1G
APOSOR917609-250MG
4-Methyl-2-oxo-2H-chromene-7-carbaldehyde 95%
4-Methyl-2-oxo-2H-chromene-7-carbaldehyde
Formula:
C₁₁H₈O₃ MW: 188,179 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00582536 CAS Number: 53183-53-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...