Benzofuran-6-carboxylic acid 98%
Supplier: Apollo Scientific
OR917818-1G
OR917818-25G
OR917818-5G
APOSOR917818-1GEA
90.2
EUR
APOSOR917818-1G
APOSOR917818-25G
APOSOR917818-5G
Benzofuran-6-carboxylic acid 98%
Benzofuran-6-carboxylic acid
Formula:
C₉H₆O₃ MW: 162,142 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01006751 CAS Number: 77095-51-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...