Ethyl 2-amino-3-(pyridin-4-yl)propanoate 95%

Supplier: Apollo Scientific

OR918142-1G OR918142-250MG
APOSOR918142-1GEA 771 EUR
APOSOR918142-1G APOSOR918142-250MG
Ethyl 2-amino-3-(pyridin-4-yl)propanoate 95%
Ethyl 2-amino-3-(pyridin-4-yl)propanoate
Formula: C₁₀H₁₄N₂O₂
MW: 194,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05228162
CAS Number: 103392-91-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR