2-Hydroxy-8-methylquinoline-3-carbaldehyde 95%
Supplier: Apollo Scientific
OR918309-1G
OR918309-250MG
APOSOR918309-1GEA
574
EUR
APOSOR918309-1G
APOSOR918309-250MG
2-Hydroxy-8-methylquinoline-3-carbaldehyde 95%
2-Hydroxy-8-methylquinoline-3-carbaldehyde
Formula:
C₁₁H₉NO₂ MW: 187,195 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02995894 CAS Number: 101382-54-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...