Dimethyl methylmalonate 95+%

Supplier: Apollo Scientific
Warning

Synonyms: Dimethyl 2-methylmalonate

OR918351-25G OR918351-5G OR918351-100G
APOSOR918351-25GEA 164 EUR
APOSOR918351-25G APOSOR918351-5G APOSOR918351-100G
Dimethyl methylmalonate 95+%
Dimethyl methylmalonate
Formula: C₆H₁₀O₄
MW: 146,141 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00075185
CAS Number: 609-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR