3-(N-Phenyl-N-methyl)aminoacrolein 98%

Supplier: Apollo Scientific

Synonyms: 3-(Methyl(phenyl)amino)acrylaldehyde

OR918466-1G OR918466-5G
APOSOR918466-1GEA 33.5 EUR
APOSOR918466-1G APOSOR918466-5G
3-(N-Phenyl-N-methyl)aminoacrolein 98%
3-(N-Phenyl-N-methyl)aminoacrolein
Formula: C₁₀H₁₁NO
MW: 161,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00190652
CAS Number: 14189-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR