Methyl 2-amino-4-methoxybenzoate 98%
Supplier: Apollo Scientific
Synonyms:
Methyl 2-amino-4-methoxylbenzoate
OR918474-1G
APOSOR918474-1GEA
0
EUR
APOSOR918474-1G
Methyl 2-amino-4-methoxybenzoate 98%
Methyl 2-amino-4-methoxybenzoate
Formula:
C₉H₁₁NO₃ MW: 181,189 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06658470 CAS Number: 50413-30-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...