N-(4-Nitrophenyl)propionamide 95%
Supplier: Apollo Scientific
OR918809-1G
OR918809-5G
APOSOR918809-1GEA
21.5
EUR
APOSOR918809-1G
APOSOR918809-5G
N-(4-Nitrophenyl)propionamide 95%
N-(4-Nitrophenyl)propionamide
Formula:
C₉H₁₀N₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD00791260 CAS Number: 4850-93-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...