2-Chloro-6-hydroxy-7-methylpurine 95%

Supplier: Apollo Scientific

Synonyms: 2-Chloro-7-methyl-7H-purin-6-ol

OR918947-1G OR918947-250MG
APOSOR918947-1GEA 353 EUR
APOSOR918947-1G APOSOR918947-250MG
2-Chloro-6-hydroxy-7-methylpurine 95%
2-Chloro-6-hydroxy-7-methylpurine
Formula: C₆H₅ClN₄O
MW: 184,583 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00047225
CAS Number: 16017-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR