3-(Prop-2-ynyloxy)pyridine 95%
Supplier: Apollo Scientific
Synonyms:
3-(Prop-2-yn-1-yloxy)pyridine
OR919014-1G
OR919014-5G
APOSOR919014-1GEA
65.4
EUR
APOSOR919014-1G
APOSOR919014-5G
3-(Prop-2-ynyloxy)pyridine 95%
3-(Prop-2-ynyloxy)pyridine
Formula:
C₈H₇NO MW: 133.147 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10698154 CAS Number: 69022-70-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...