Adenine hydrochloride hydrate 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
6-Aminopurine hydrochloride hydrate
OR919168-100G
OR919168-500G
APOSOR919168-100GEA
75.4
EUR
APOSOR919168-100G
APOSOR919168-500G
Adenine hydrochloride hydrate 95%
Adenine hydrochloride hydrate
Formula:
C₅H₅N₅·HCl·xH₂O MW: 171,59 g/mol (anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00150864 CAS Number: 916986-40-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...