3-(4-Chlorophenyl)prop-2-ynamide 95%

Supplier: Apollo Scientific

OR919578-1G OR919578-250MG
APOSOR919578-1GEA 864 EUR
APOSOR919578-1G APOSOR919578-250MG
3-(4-Chlorophenyl)prop-2-ynamide 95%
3-(4-Chlorophenyl)prop-2-ynamide
Formula: C₉H₆ClNO
Storage Temperature: Ambient
MDL Number: MFCD20699326
CAS Number: 42122-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR