6-Bromo-3-hydroxypyrazine-2-carboxamide

Supplier: Apollo Scientific

OR919629-1G OR919629-250MG OR919629-5G
APOSOR919629-1GEA 221 EUR
APOSOR919629-1G APOSOR919629-250MG APOSOR919629-5G
6-Bromo-3-hydroxypyrazine-2-carboxamide
6-Bromo-3-hydroxypyrazine-2-carboxamide
Formula: C₅H₄BrN₃O₂
MW: 218,008 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20528005
CAS Number: 259793-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR