Ethyl 7-chloroimidazo[1,2-a]pyridine-3-carboxylate 97%

Supplier: Apollo Scientific
Warning

OR919720-1G OR919720-5G
APOSOR919720-1GEA 430 EUR
APOSOR919720-1G APOSOR919720-5G
Ethyl 7-chloroimidazo[1,2-a]pyridine-3-carboxylate 97%
Ethyl 7-chloroimidazo[1,2-a]pyridine-3-carboxylate
Formula: C₁₀H₉ClN₂O₂
MW: 224,644 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22200342
CAS Number: 1296201-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR