(2S)-2-Isopropylpyrrolidine 95%
Supplier: Apollo Scientific
OR921148-100MG
OR921148-1G
OR921148-250MG
APOSOR921148-100MGEA
190
EUR
APOSOR921148-100MG
APOSOR921148-1G
APOSOR921148-250MG
(2S)-2-Isopropylpyrrolidine 95%
(2S)-2-Isopropylpyrrolidine
Formula:
C₇H₁₅N MW: 113,201 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10696887 CAS Number: 41720-99-4 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...