4-Ethynyltetrahydropyran-4-ol 98%
Supplier: Apollo Scientific
OR921326-1G
OR921326-250MG
APOSOR921326-1GEA
632
EUR
APOSOR921326-1G
APOSOR921326-250MG
4-Ethynyltetrahydropyran-4-ol 98%
4-Ethynyltetrahydropyran-4-ol
Formula:
C₇H₁₀O₂ Storage Temperature: Ambient |
MDL Number:
MFCD18814953 CAS Number: 57385-16-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...