3-Nitro-6-pyridinecarboxaldehyde 95%
Supplier: Apollo Scientific
Synonyms:
5-Nitropicolinaldehyde
OR921887-1G
OR921887-5G
APOSOR921887-1GEA
156
EUR
APOSOR921887-1G
APOSOR921887-5G
3-Nitro-6-pyridinecarboxaldehyde 95%
3-Nitro-6-pyridinecarboxaldehyde
Formula:
C₆H₄N₂O₃ MW: 152,108 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07368185 CAS Number: 35969-75-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...