2-(2-Methylphenyl)pyrimidine-5-carbaldehyde 95%
Supplier: Apollo Scientific
OR921955-1G
OR921955-250MG
APOSOR921955-1GEA
400
EUR
APOSOR921955-1G
APOSOR921955-250MG
2-(2-Methylphenyl)pyrimidine-5-carbaldehyde 95%
2-(2-Methylphenyl)pyrimidine-5-carbaldehyde
Formula:
C₁₂H₁₀N₂O MW: 198,221 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07643179 CAS Number: 876710-73-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...