1-Benzyl-4-methyl-1,2,3,6-tetrahydropyridine ≥98%

Supplier: Apollo Scientific

OR922045-1G OR922045-5G
APOSOR922045-1GEA 133 EUR
APOSOR922045-1G APOSOR922045-5G
1-Benzyl-4-methyl-1,2,3,6-tetrahydropyridine ≥98%
1-Benzyl-4-methyl-1,2,3,6-tetrahydropyridine
Formula: C₁₃H₁₇N
MW: 187,281 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09030042
CAS Number: 32018-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR