N-(2-Cyano-cyclohex-1-enyl)-acetamide 95%
Supplier: Apollo Scientific
OR922083-1G
OR922083-250MG
APOSOR922083-1GEA
437
EUR
APOSOR922083-1G
APOSOR922083-250MG
N-(2-Cyano-cyclohex-1-enyl)-acetamide 95%
N-(2-Cyano-cyclohex-1-enyl)-acetamide
Formula:
C₉H₁₂N₂O MW: 164,204 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00102302 CAS Number: 500896-58-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...