5-Methyl-2-phenyl-3-furoic acid 97%

Supplier: Apollo Scientific

OR9223-1G OR9223-250MG
APOSOR9223-1GEA 176 EUR
APOSOR9223-1G APOSOR9223-250MG
5-Methyl-2-phenyl-3-furoic acid 97%
5-Methyl-2-phenyl-3-furoic acid
Formula: C₁₂H₁₀O₃
MW: 202.206 g/mol
Melting Pt: 145…147.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00447900
CAS Number: 64354-50-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR