2-Hydroxy-N-methylbenzamide 95%
Supplier: Apollo Scientific
Synonyms:
N-Methylsalicylamide
OR922451-5G
APOSOR922451-5GEA
177
EUR
APOSOR922451-5G
2-Hydroxy-N-methylbenzamide 95%
2-Hydroxy-N-methylbenzamide
Formula:
C₈H₉NO₂ MW: 151,163 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00045757 CAS Number: 1862-88-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...