2-(Hydroxymethyl)cyclobutan-1-one
Supplier: Apollo Scientific
Synonyms:
2-(Hydroxymethyl)cyclobutanone
OR922566-1G
APOSOR922566-1GEA
683
EUR
APOSOR922566-1G
2-(Hydroxymethyl)cyclobutan-1-one
2-(Hydroxymethyl)cyclobutan-1-one
Formula:
C₅H₈O₂ MW: 100,116 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD23106469 CAS Number: 23107-52-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...