3,4,5-Trichloropyridin-2-amine 95%
Supplier: Apollo Scientific
Synonyms:
2-Amino-3,4,5-trichloropyridine
OR922583-1G
APOSOR922583-1GEA
72.5
EUR
APOSOR922583-1G
3,4,5-Trichloropyridin-2-amine 95%
3,4,5-Trichloropyridin-2-amine
Formula:
C₅H₃Cl₃N₂ MW: 197,45 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08705655 CAS Number: 55933-91-0 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...