tert-Butyl ethyl oxalate 95%

Supplier: Apollo Scientific

OR922614-5G
APOSOR922614-5GEA 194 EUR
APOSOR922614-5G
tert-Butyl ethyl oxalate 95%
tert-Butyl ethyl oxalate
Formula: C₈H₁₄O₄
MW: 174,194 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11052953
CAS Number: 50624-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR