Benzyl 3-methyl-4-oxopiperidine-1-carboxylate

Supplier: Apollo Scientific
Warning

OR92275-1G OR92275-5G
APOSOR92275-1GEA 126 EUR
APOSOR92275-1G APOSOR92275-5G
Benzyl 3-methyl-4-oxopiperidine-1-carboxylate
Benzyl 3-methyl-4-oxopiperidine-1-carboxylate
Formula: C₁₄H₁₇NO₃
MW: 247,29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11848402
CAS Number: 1010115-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR