4-Methylthiophen-3-amine 95%

Supplier: Apollo Scientific

OR922825-100MG OR922825-1G OR922825-250MG
APOSOR922825-100MGEA 168 EUR
APOSOR922825-100MG APOSOR922825-1G APOSOR922825-250MG
4-Methylthiophen-3-amine 95%
4-Methylthiophen-3-amine
Formula: C₅H₇NS
MW: 113,181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09907775
CAS Number: 23967-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR