2-(2H-1,2,3-Triazol-2-yl)benzoic acid 95%
Supplier: Apollo Scientific
OR923048-1G
OR923048-5G
APOSOR923048-1GEA
77
EUR
APOSOR923048-1G
APOSOR923048-5G
2-(2H-1,2,3-Triazol-2-yl)benzoic acid 95%
2-(2H-1,2,3-Triazol-2-yl)benzoic acid
Formula:
C₉H₇N₃O₂ MW: 189,171 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20486491 CAS Number: 1001401-62-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...