Ethyl cis-2-aminocyclopentanecarboxylate 95%
Supplier: Apollo Scientific
OR923247-1G
OR923247-250MG
APOSOR923247-1GEA
720
EUR
APOSOR923247-1G
APOSOR923247-250MG
Ethyl cis-2-aminocyclopentanecarboxylate 95%
Ethyl cis-2-aminocyclopentanecarboxylate
Formula:
C₈H₁₅NO₂ MW: 157,21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09475430 CAS Number: 114745-45-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...