5-Methyl-2-phenyl-1,3-oxazole-4-carboxaldehyde 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(4-Formyl-5-methyl-1,3-oxazol-2-yl)benzene, 4-Formyl-5-methyl-2-phenyl-1,3-oxazole
OR9233-1G
OR9233-250MG
APOSOR9233-1GEA
263
EUR
APOSOR9233-1G
APOSOR9233-250MG
5-Methyl-2-phenyl-1,3-oxazole-4-carboxaldehyde 97%
5-Methyl-2-phenyl-1,3-oxazole-4-carboxaldehyde
Formula:
C₁₁H₉NO₂ MW: 187,195 g/mol Melting Pt: 87…88 °C Storage Temperature: Ambient |
MDL Number:
MFCD08435848 CAS Number: 70170-23-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...