5-Amino-2,3-dimethyl-2H-indazole 95%
Supplier: Apollo Scientific
OR923392-1G
OR923392-250MG
APOSOR923392-1GEA
807
EUR
APOSOR923392-1G
APOSOR923392-250MG
5-Amino-2,3-dimethyl-2H-indazole 95%
5-Amino-2,3-dimethyl-2H-indazole
Formula:
C₉H₁₁N₃ MW: 161,204 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD17015994 CAS Number: 221681-94-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...