Methyl 2-cyclopropylacetate 95%

Supplier: Apollo Scientific

OR923404-1G OR923404-5G
APOSOR923404-1GEA 79.9 EUR
APOSOR923404-1G APOSOR923404-5G
Methyl 2-cyclopropylacetate 95%
Methyl 2-cyclopropylacetate
Formula: C₆H₁₀O₂
MW: 114,142 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12031699
CAS Number: 34108-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR