2-Amino-3-(morpholinomethyl)pyridine 95%

Supplier: Apollo Scientific

OR923473-1G OR923473-250MG
APOSOR923473-1GEA 790 EUR
APOSOR923473-1G APOSOR923473-250MG
2-Amino-3-(morpholinomethyl)pyridine 95%
2-Amino-3-(morpholinomethyl)pyridine
Formula: C₁₀H₁₅N₃O
MW: 193,246 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16765022
CAS Number: 1250814-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR