5-Chlorothieno[3,2-b]pyridine 95%
Supplier: Apollo Scientific
OR923512-1G
OR923512-250MG
OR923512-5G
APOSOR923512-1GEA
263
EUR
APOSOR923512-1G
APOSOR923512-250MG
APOSOR923512-5G
5-Chlorothieno[3,2-b]pyridine 95%
5-Chlorothieno[3,2-b]pyridine
Formula:
C₇H₄ClNS MW: 169,631 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09909817 CAS Number: 65977-55-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...