4-Chloro-5,6-dimethylpyrimidine 95%
Supplier: Apollo Scientific
OR923520-1G
OR923520-250MG
APOSOR923520-1GEA
793
EUR
APOSOR923520-1G
APOSOR923520-250MG
4-Chloro-5,6-dimethylpyrimidine 95%
4-Chloro-5,6-dimethylpyrimidine
Formula:
C₆H₇ClN₂ MW: 142,586 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09909775 CAS Number: 67434-65-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...