tert-Butyl l-valinate 96%

Supplier: Apollo Scientific

Synonyms: (S)-tert-Butyl 2-amino-3-methylbutanoate, H-Val-OtBu

OR923559-25G OR923559-5G OR923559-100G
APOSOR923559-25GEA 180 EUR
APOSOR923559-25G APOSOR923559-5G APOSOR923559-100G
tert-Butyl l-valinate 96%
tert-Butyl l-valinate
Formula: C₉H₁₉NO₂
MW: 173.253 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00056147
CAS Number: 13211-31-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR