Methyl 3-aminopyridazine-6-carboxylate hydrochloride 95%

Supplier: Apollo Scientific

OR923653-1G OR923653-250MG
APOSOR923653-1GEA 51.5 EUR
APOSOR923653-1G APOSOR923653-250MG
Methyl 3-aminopyridazine-6-carboxylate hydrochloride 95%
Methyl 3-aminopyridazine-6-carboxylate hydrochloride
Formula: C₆H₇N₃O₂·ClH
Storage Temperature: Ambient
MDL Number: MFCD17016033
CAS Number: 98140-96-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR