1,2,3,4-Tetrahydroquinolin-3-ol 96%
Supplier: Apollo Scientific
OR924505-1G
OR924505-250MG
APOSOR924505-1GEA
329
EUR
APOSOR924505-1G
APOSOR924505-250MG
1,2,3,4-Tetrahydroquinolin-3-ol 96%
1,2,3,4-Tetrahydroquinolin-3-ol
Formula:
C₉H₁₁NO MW: 149,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11616169 CAS Number: 3418-45-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...