2,2-Dimethylbenzofuran-3(2H)-one 95%
Supplier: Apollo Scientific
OR924544-1G
OR924544-250MG
APOSOR924544-1GEA
719
EUR
APOSOR924544-1G
APOSOR924544-250MG
2,2-Dimethylbenzofuran-3(2H)-one 95%
2,2-Dimethylbenzofuran-3(2H)-one
Formula:
C₁₀H₁₀O₂ MW: 162,185 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20040044 CAS Number: 16748-90-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...