5,8-Dioxaspiro[3.4]octan-2-ylmethanamine

Supplier: Apollo Scientific

OR924669-1G OR924669-5G
APOSOR924669-1GEA 432 EUR
APOSOR924669-1G APOSOR924669-5G
5,8-Dioxaspiro[3.4]octan-2-ylmethanamine
5,8-Dioxaspiro[3.4]octan-2-ylmethanamine
Formula: C₇H₁₃NO₂
MW: 143,184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23106176
CAS Number: 1392804-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR