5-Methylbenzofuran 95%
Supplier: Apollo Scientific
OR924946-1G
OR924946-250MG
APOSOR924946-1GEA
719
EUR
APOSOR924946-1G
APOSOR924946-250MG
5-Methylbenzofuran 95%
5-Methylbenzofuran
Formula:
C₉H₈O Storage Temperature: Ambient |
MDL Number:
MFCD00138124 CAS Number: 18441-43-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...