6-Methyl-3,5-heptadien-2-one 95%

Supplier: Apollo Scientific

Synonyms: 6-Methylhepta-3,5-dien-2-one

OR925267-25G
APOSOR925267-25GEA 104 EUR
APOSOR925267-25G
6-Methyl-3,5-heptadien-2-one 95%
6-Methyl-3,5-heptadien-2-one
Formula: C₈H₁₂O
MW: 124,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00043647
CAS Number: 16647-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR