2-Bromopyrimidin-5-ol 95%
Supplier: Apollo Scientific
Synonyms:
2-Bromo-5-hydroxypyrimidine
OR925447-1G
OR925447-250MG
APOSOR925447-1GEA
466
EUR
APOSOR925447-1G
APOSOR925447-250MG
2-Bromopyrimidin-5-ol 95%
2-Bromopyrimidin-5-ol
Formula:
C₄H₃BrN₂O MW: 174,983 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09953508 CAS Number: 1240621-87-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...