4-Phenyl-1,3-thiazole-2-carbaldehyde 95%

Supplier: Apollo Scientific
Warning

OR925848-1G OR925848-250MG
APOSOR925848-1GEA 185 EUR
APOSOR925848-1G APOSOR925848-250MG
4-Phenyl-1,3-thiazole-2-carbaldehyde 95%
4-Phenyl-1,3-thiazole-2-carbaldehyde
Formula: C₁₀H₇NOS
Storage Temperature: Ambient
MDL Number: MFCD02665589
CAS Number: 75390-44-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR