(2E)-3-(1,5-Dimethyl-1H-pyrazol-4-yl)acrylic acid 95%
Supplier: Apollo Scientific
OR925918-1G
OR925918-250MG
APOSOR925918-1GEA
720
EUR
APOSOR925918-1G
APOSOR925918-250MG
(2E)-3-(1,5-Dimethyl-1H-pyrazol-4-yl)acrylic acid 95%
(2E)-3-(1,5-Dimethyl-1H-pyrazol-4-yl)acrylic acid
Formula:
C₈H₁₀N₂O₂ MW: 166,177 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02090861 CAS Number: 689251-96-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...