3-Nitropicolinaldehyde 90%

Supplier: Apollo Scientific

Synonyms: 3-Nitro-2-pyridinecarboxaldehyde

OR926080-1G OR926080-250MG OR926080-5G
APOSOR926080-1GEA 132 EUR
APOSOR926080-1G APOSOR926080-250MG APOSOR926080-5G
3-Nitropicolinaldehyde 90%
3-Nitropicolinaldehyde
Formula: C₆H₄N₂O₃
MW: 152,108 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10696871
CAS Number: 10261-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR